Acetone 99+%, extra pure
Supplier: Thermo Fisher Scientific
Acetone 99+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56 °C (760 mmHg)
- Melting Pt:–95 °C
- Density:0.79 g/cm³
- Flash Pt:–20 °C
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
5 Options Available Below
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56 °C (760 mmHg)
- Melting Pt:–95 °C
- Density:0.79 g/cm³
- Flash Pt:–20 °C
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.5 % (K.F.)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
Specifications
Specifications
- Catalog No:
- ACRO177170010
- ACRO177170025
- ACRO177170050
- ACRO177170100
- ACRO177170250
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.ACRO177170010Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.ACRO177170025Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.ACRO177170050Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.ACRO177170100Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health - Specifications:
Safer Human and Environmental Health More
Specifications
Cat. No.ACRO177170250Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



