Order Entry
Germany
ContactUsLinkComponent
Methyl ethyl ketone, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Methyl ethyl ketone, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
undefined
Methyl ethyl ketone, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Solubility:
    292 g/l (20 °C)
  • Melting Point:
    -86 °C
  • Molar Mass:
    72.11 g/mol
  • Boiling Point:
    79.6 °C (1013 hPa)
  • Vapor Pressure:
    105 hPa (20 °C)
  • Flash Point:
    -4 °C
  • Refractive Index:
    1.3814 (15 °C)
  • Explosion Limit:
    1.8 - 11.5 %(V)
  • Density:
    0.805 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) neutral
  • Ignition Point:
    514 °C
  • Evaporation number:
    6.3

Specification Test Results

Related Information

Certificates
Search by Lot Number