

Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-93-3
:
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648
Merck Index:13,06097
Boiling Pt:79.6 °C (1013 hPa)
Melting Pt:–87 °C
Density:0.806 g/cm³
Flash Pt:–7 °C
Storage Temperature:Ambient
UN:1193
ADR:3,II
:72.11 g/mol
EINECS:201-159-0
REACH:01-2119457290-43
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.1 %
- GC:≥99.0 %
- UV:1 cm cell vs water
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %