Triethylammonium chloride, Sigma-Aldrich®
: Merck
:554-68-7
:
:C₆H₁₆ClN
:MFCD00012500
:137.65 g/mol
:0.7637 g/cm³ (20 °C)
:Ambient
:209-067-2
:1759
:8,III
:M-Clarity™ quality level = MQ200
:GHS05
- Solubility:1440 g/l (20 °C)
- Melting Point:255 - 258 °C
- Molar Mass:137.65 g/mol
- Bulk Density:220 kg/m3
- Density:1.07 g/cm3 (21 °C)
- pH:5 (10 g/l, H2O, 20 °C)
- Synonyms:Triethylamine hydrochloride
- Hill Formula:C6H16ClN
- Chemical Formula:(C2H5)3N * HCl