0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
trans-4-Aminocyclohexanol hydrochloride
trans-4-Aminocyclohexanol hydrochloride 97%
trans-4-Aminocyclohexanol hydrochloride 97%
:
Thermo Fisher Scientific
trans-4-Aminocyclohexanol hydrochloride 97%
:
Thermo Fisher Scientific
:
50910-54-8
:
:
C₆H₁₄ClNO
:
MFCD00012566
:
151.64 g/mol
:
225…230 °C
:
GHS07
:
Ambient
msds_ALFAB25070
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years