Order Entry
Germany
ContactUsLinkComponent
Organic reference standard, Tetrachloromethane 1,000 µg/ml in methanol
Organic reference standard, Tetrachloromethane 1,000 µg/ml in methanol
Catalog # CMPLE798.1K.ML.1
CAS Number:  
Organic reference standard, Tetrachloromethane 1,000 µg/ml in methanol
Catalog # CMPLE798.1K.ML.1
CAS Number:  

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    1 Ampoul
  • CAS No.:
    56-23-5
  • Description:
    Tetrachloromethane 1,000 µg/ml in methanol
  • Formula:
    CCl₄
  • Description:
    Tetrachloromethane

Specifications

About this item