Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium hydroxide
Sodium hydroxide 33% in aqueous solution
Sodium hydroxide 33% in aqueous solution
Sodium hydroxide 33% in aqueous solution
OVER813239
:
OVERLACK CHEMISCHE PRODUKTE
:
Sodium hydroxide 33% in aqueous solution
OVER813239
:
OVERLACK CHEMISCHE PRODUKTE
:
Pk:
1.000 kg
Seq:
4876
:
:
40 g/mol
:
119…135 °C (1013 hPa)
:
9…10 °C
:
1,53 g/cm³ (20 °C)
:
Ambient
:
215-185-5
:
1824
:
8,II
:
GHS05
:
1310-73-2
:
NaOH
:
MFCD00003548