

Butyl methacrylate 99% stabilised
:97-88-1
:
:C₈H₁₄O₂
:MFCD00009444
:163 °C (1013 hPa)
:–75 °C
:0.89 g/cm³
:41 °C
:Refrigerator
:202-615-1
:3,III
:142.2 g/mol
:2227
:GHS02
:GHS07
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4225 to 1.4245 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤10 APHA
- Stabilizer:10 to 110 ppm MEHQ