Order Entry
Germany
ContactUsLinkComponent
 
 
 
 
Methylmagnesium bromide 3 M in diethyl ether, AcroSeal®
 
undefined
 
 
 
Methylmagnesium bromide 3 M in diethyl ether, AcroSeal®

 

  • Appearance:
    Clear to hazy grey to brown to black Solution (May develop some turbidity or precipitate)
  • Titration Grignard:
    2.85 to 3.20 M
  • Melting point:
    120 to 124 °C
  • Solubility:
    (5 % in ethanol) Clear solution
  • Residue after ignition:
    ≤0.1 %
  • Titration with HClO4:
    97.5 to 102.5 %