Order Entry
Germany
ContactUsLinkComponent
Methylmagnesium bromide 3 mol/l in diethyl ether, Sigma-Aldrich®
Methylmagnesium bromide 3 mol/l in diethyl ether, Sigma-Aldrich®
Catalog # SIAL257087-200ML
Supplier:  Merck
CAS Number:  
Methylmagnesium bromide 3 mol/l in diethyl ether, Sigma-Aldrich®
Catalog # SIAL257087-200ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    200 mL
  • Seq:
    2530

Specifications

About this item

 :
 :119.24 g/mol
ADR:4.3,I
 :M-Clarity™ quality level = MQ100
EINECS:200-844-1
UN:1928
CAS Number:75-16-1
Formula:CH₃BrMg
MDL Number:MFCD00000041