
Heptafluorobutyric acid
Catalog # PIER25003
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
Boiling Pt:121 °C (1013 hPa)
Melting Pt:–17.5 °C
Density:1.764 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:8,II
:214.04 g/mol
EINECS:206-786-3
UN:3265
:GHS05
CAS Number:375-22-4
Formula:C₄HF₇O₂
MDL Number:MFCD00004171