

Diethyl ether ≥99.5% (by GC) stabilised, Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:34…35 °C (1.013 hPa)
Melting Pt:–116 °C (1.013 hPa)
Density:0.71 g/cm³ (20 °C)
Flash Pt:–45 °C (1.013 hPa)
Storage Temperature:Ambient
UN:1155
ADR:3,I
:Stabilised with 2,6-Di-tert-butyl-4-methylphenol ~5 mg/L
EINECS:200-467-2
REACH:01-2119535785-29
:GHS02
:GHS07
Specification Test Results
- Refractive Index:n20/D 1.3530 (lit.)
- Identity A,B:complying
- Assay (GC):Min. 99.5 %
- Boiling range:34,0 - 35,0 °C
- Density (D 20/20):0.714 - 0.716
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.2 %
- Free acid (as CH₃COOH):Max. 0.002 %
- Aldehydes:complying
- Peroxides (as H₂O₂):Max. 0.0002 %
- Acidity:complying
- Assay BHT (GC):5 - 8 ppm
- Residual solvents:complying
- Vapor Density:2.6 (vs air)