Order Entry
Germany
ContactUsLinkComponent
Acetic acid 99-100%, Puriss., meets analytical specification of BP, Ph. Eur., USP
Acetic acid 99-100%, Puriss., meets analytical specification of BP, Ph. Eur., USP
Certificates
undefined
Acetic acid 99-100%, Puriss., meets analytical specification of BP, Ph. Eur., USP
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Non-volatile matter:
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.0005 %
  • Heavy metals (as Pb):
    Max. 0.0005 %
  • Chloride (Cl):
    Max. 0.0005 %
  • Sulfate (SO₄):
    Max. 0.0005 %
  • Explosion Limit:
    16 %, 92 °F
  • Vapor Density:
    2.07 (vs air)
  • Autoignition Temperature:
    800 °F

Specification Test Results

Related Information