Digitoxin
Glass bottle for solids, 100 mg
Catalog # PROOICRS0288
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:71-63-6
- Formula:C₄₁H₆₄O₁₃
- MDL Number:MFCD00003686
- Molecular Weight:764.95 g/mol
- Melting Pt:276 °C
- EINECS:200-760-5
- UN:2811
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



