Order Entry
Germany
ContactUsLinkComponent
Cytochalasin B (from Helminthosporium dematioideum), Millipore®
Cytochalasin B (from Helminthosporium dematioideum), Millipore®
Catalog # 250233-5
CAS Number:  
Cytochalasin B (from Helminthosporium dematioideum), Millipore®
Catalog # 250233-5
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :479.62 g/mol
EINECS:239-000-2
UN:2811
ADR:6.1,II
CAS Number:14930-96-2
Formula:C₂₉H₃₇NO₅
MDL Number:MFCD00077704