
cis-2-Aminocyclohexanol hydrochloride 99+%
:6936-47-6
:
:C₆H₁₄ClNO
:MFCD00143981
:151.64 g/mol
:186…190 °C
:GHS07
:Ambient
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:186 to 190 °C
- Titration with HClO4:≥99.0 %