Ammonium hexachloroosmate(IV), Sigma-Aldrich®
SIAL544922-1G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
:
:170 °C
:2.93 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:439.02 g/mol
:235-188-5
:12125-08-5
:H₈Cl₆N₂Os
:MFCD00010883