Order Entry
Germany
ContactUsLinkComponent
Ammonium hexachloroosmate(IV) 99.99% (trace metals basis)
Irritant
Ammonium hexachloroosmate(IV) 99.99% (trace metals basis)
Catalog # ACRO197630010
CAS Number:  
undefined
Irritant
Ammonium hexachloroosmate(IV) 99.99% (trace metals basis)
Catalog # ACRO197630010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Red to black Powder
  • Total trace metal impurities:
    ≤100 ppm

Specification Test Results

About this item

 :
Melting Pt:170 °C
Density:2,93 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,III
 :439.02 g/mol
EINECS:235-188-5
 :GHS07
CAS Number:12125-08-5
Formula:H₈Cl₆N₂Os
MDL Number:MFCD00010883