Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
9-Bromoanthracene
9-Bromoanthracene
9-Bromoanthracene
9-Bromoanthracene
Supplier:
COMBI-BLOCKS
Certificates
Expand image
9-Bromoanthracene
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
1564-64-3
:
Formula:
C₁₄H₉Br
MDL Number:
MFCD00001243
:
257.13 g/mol
Boiling Pt:
359 °C (1013 hPa)
Melting Pt:
103 °C
Storage Temperature:
Ambient
EINECS:
216-359-3