7-Methoxyindole
Supplier: COMBI-BLOCKS
Certificates
About this item
CAS Number:3189-22-8
:
Formula:C₉H₉NO
MDL Number:MFCD00047203
:147.18 g/mol
Boiling Pt:298 °C (1013 hPa)
Density:1,126 g/cm³ (25 °C)
EINECS:221-690-1
:GHS07
Storage Temperature:Freezer