0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
2-Chloro-5-pyridineboronic acid
2-Chloro-5-pyridineboronic acid
2-Chloro-5-pyridineboronic acid
2-Chloro-5-pyridineboronic acid
Supplier:
COMBI-BLOCKS
Certificates
Expand image
2-Chloro-5-pyridineboronic acid
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
444120-91-6
:
Formula:
C₅H₅BClNO₂
MDL Number:
MFCD03094998
:
157.36 g/mol