Methyl-4-chlorobenzoate
Supplier: COMBI-BLOCKS
Certificates
About this item
CAS Number:1126-46-1
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00000621
:170.6 g/mol
Merck Index:13,02144
Boiling Pt:103…106 °C (15 mmHg)
Melting Pt:43 °C
Density:1.382 g/cm³ (20 °C)
EINECS:214-420-9
Frequently Bought Together