(±)-3-Methyl-3-hexanol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:597-96-6
:
Formula:C₇H₁₆O
MDL Number:MFCD00021839
:116.2 g/mol
Boiling Pt:143 °C
Density:0.823
Flash Pt:45 °C (113 °F)
Storage Temperature:Ambient
EINECS:209-910-4
UN:1987
ADR:3,III
:GHS02
:GHS07