


2-Mercaptoethanol
Catalog # BC98
Supplier: G-Biosciences
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
Merck Index:13,05894
Boiling Pt:157 °C (1013 hPa)
Melting Pt:–100 °C
Density:1.12 g/cm³ (20 °C)
Flash Pt:68 °C
Storage Temperature:Ambient
EINECS:200-464-6
UN:2966
ADR:6.1,II
:78.14 g/mol
:GHS06
:GHS05
:GHS09
CAS Number:60-24-2
Formula:HSCH₂CH₂OH
MDL Number:MFCD00004890