2-Iodobenzoic acid
Supplier: COMBI-BLOCKS
Certificates
About this item
CAS Number:88-67-5
:
Formula:C₇H₅IO₂
MDL Number:MFCD00002419
:248.02 g/mol
Merck Index:13,05052
Boiling Pt:314 °C (1013 hPa)
Melting Pt:159…160 °C
Density:2.25 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-850-7
Frequently Bought Together