2-Hydroxy-5-nitropyridine
Marketsource
Supplier: COMBI-BLOCKS
Jump to:
About this item
Synonyms: 5-Nitro-2-pyridinol
- CAS Number:5418-51-9
- Formula:C₅H₄N₂O₃
- MDL Number:MFCD00006276
- Molecular Weight:140.1 g/mol
- Boiling Pt:180…200 °C (0,2 mmHg)
- Boiling Pt:180…200 °C (0.2 mmHg)
- Melting Pt:191…192 °C
- EINECS:226-525-7
3 Options Available Below
Product Details & Documents
Synonyms: 5-Nitro-2-pyridinol
- CAS Number:5418-51-9
- Formula:C₅H₄N₂O₃
- MDL Number:MFCD00006276
- Molecular Weight:140.1 g/mol
- Boiling Pt:180…200 °C (0,2 mmHg)
- Boiling Pt:180…200 °C (0.2 mmHg)
- Melting Pt:191…192 °C
- EINECS:226-525-7
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



