2-Ethylhexane-1,3-diol (mixture of isomers), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:94-96-2
:
Formula:C₈H₁₈O₂
MDL Number:MFCD00004578
:146.23 g/mol
Boiling Pt:244 °C (1013 hPa)
Melting Pt:–40 °C
Density:0.9422 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:202-377-9