Order Entry
Germany
ContactUsLinkComponent
 
2-Chloro-5-(chloromethyl)pyridine
Catalog # EHERC11397000
Supplier:  EHRENSTORFER
CAS Number:  
2-Chloro-5-(chloromethyl)pyridine
Catalog # EHERC11397000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :162.02 g/mol
Melting Pt:37 °C
UN:3261
ADR:8,II
CAS Number:70258-18-3
Formula:C₆H₅Cl₂N
MDL Number:MFCD00125366