Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4-(trifluoromethyl)phenylboronic acid
2-Chloro-4-(trifluoromethyl)phenylboronic acid
2-Chloro-4-(trifluoromethyl)phenylboronic acid
2-Chloro-4-(trifluoromethyl)phenylboronic acid
Supplier:
COMBI-BLOCKS
Certificates
Expand image
2-Chloro-4-(trifluoromethyl)phenylboronic acid
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
254993-59-4
:
Formula:
C₇H₅BClF₃O₂
MDL Number:
MFCD02684337
:
224.37