
2-Isopropylimidazole ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36947-68-9
:
Formula:C₆H₁₀N₂
MDL Number:MFCD00014486
:110.16 g/mol
Boiling Pt:256…260 °C
Melting Pt:126…132 °C
Storage Temperature:Ambient
EINECS:253-286-6
:GHS07