2-(2-Aminoethyl)pyridine
Supplier: CHEMPUR
Certificates
About this item
CAS Number:2706-56-1
:
Formula:C₇H₁₀N₂
MDL Number:MFCD00006367
:122.17 g/mol
Boiling Pt:49…50 °C (0,03 mmHg)
Density:1,02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:220-295-1