1-(2-Pyridylazo)-2-naphthol (PAN)
Catalog # MOLE63973572-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:249.27 g/mol
Boiling Pt:412 °C (1013 hPa)
Melting Pt:137…140 °C
Storage Temperature:Ambient
EINECS:201-637-9
CAS Number:85-85-8
Formula:C₁₅H₁₁N₃O
MDL Number:MFCD00004071