Order Entry
Germany
ContactUsLinkComponent
 
Pesticide reference standard, 2,4,6-Trichlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA17774600ME
 :  
Pesticide reference standard, 2,4,6-Trichlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
  EHERXA17774600ME
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    88-06-2
  • Description:
    2,4,6-Trichlorophenol 100 µg/ml in methanol
  • Formula:
    C₆H₃Cl₃O
  • Description:
    2,4,6-Trichlorophenol