Pesticide reference standard, 2,4,6-Trichlorophenol 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERL17774600CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:88-06-2
- Description:2,4,6-Trichlorophenol 10 µg/ml in cyclohexane
- Formula:C₆H₃Cl₃O
- Description:2,4,6-Trichlorophenol