Pesticide reference standard, 2,4,5-Trichlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA17774500ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:95-95-4
- Description:2,4,5-Trichlorophenol 100 µg/ml in methanol
- Formula:C₆H₃Cl₃O
- Description:2,4,5-Trichlorophenol