Order Entry
Germany
ContactUsLinkComponent
Pesticide reference standard, 2,3,6-Trichlorophenol 10 µg/ml in methanol, Dr Ehrenstorfer
Pesticide reference standard, 2,3,6-Trichlorophenol 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL17773600ME
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 2,3,6-Trichlorophenol 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL17773600ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    933-75-5
  • Description:
    2,3,6-Trichlorophenol 10 µg/ml in methanol
  • Formula:
    C₆H₃Cl₃O
  • Description:
    2,3,6-Trichlorophenol

Specifications

About this item