Pesticide reference standard, 4-Chlorophenol 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA11472000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:106-48-9
- Description:4-Chlorophenol 100 µg/ml in methanol
- Formula:C₆H₅ClO
- Description:4-Chlorophenol