4-Bromo-3-fluorobenzonitrile
Catalog # FLUO034816-25G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:200.01 g/mol
UN:2811
ADR:6.1,III
CAS Number:133059-44-6
Formula:C₇H₃BrFN
MDL Number:MFCD07782069