2-(4-Bromophenyl)ethanol
Marketsource
Supplier: COMBI-BLOCKS
Jump to:
About this item
Synonyms: 4-Bromophenethyl alcohol
- CAS Number:4654-39-1
- Formula:C₈H₉BrO
- MDL Number:MFCD00002897
- Molecular Weight:201.06 g/mol
- Boiling Pt:275 °C (1013 hPa)
- Melting Pt:36…38 °C
- Density:1,4678 g/cm³ (28.5 °C)
- EINECS:225-093-7
3 Options Available Below
Product Details & Documents
Synonyms: 4-Bromophenethyl alcohol
- CAS Number:4654-39-1
- Formula:C₈H₉BrO
- MDL Number:MFCD00002897
- Molecular Weight:201.06 g/mol
- Boiling Pt:275 °C (1013 hPa)
- Melting Pt:36…38 °C
- Density:1,4678 g/cm³ (28.5 °C)
- EINECS:225-093-7
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



