Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromophenetole
3-Bromophenetole
3-Bromophenetole
3-Bromophenetole
Supplier:
COMBI-BLOCKS
Certificates
Expand image
3-Bromophenetole
Supplier:
COMBI-BLOCKS
About this item
CAS Number:
2655-84-7
:
Formula:
C₈H₉BrO
MDL Number:
MFCD00274287
:
201.06 g/mol
EINECS:
220-186-9