5'-Bromo-2'-hydroxyacetophenone
Marketsource
Supplier: CHEMPUR
Jump to:
About this item
Synonyms: 1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone
- CAS Number:1450-75-5
- Formula:C₈H₇BrO₂
- MDL Number:MFCD00191850
- Molecular Weight:215.05
2 Options Available Below
Product Details & Documents
Synonyms: 1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone
- CAS Number:1450-75-5
- Formula:C₈H₇BrO₂
- MDL Number:MFCD00191850
- Molecular Weight:215.05
Product Family Options
Product Information
- PkAvailabilityPrice
- Specifications:MarketsourceMarketsource



