Amantadine hydrochloride
Catalog # PROOBP530
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:200 mg
- Seq:2530
Specifications
About this item
:
:187.71 g/mol
Melting Pt:360 °C
EINECS:211-560-2
CAS Number:665-66-7
Formula:C₁₀H₁₈ClN
MDL Number:MFCD00074723