4-Bromo-3-(trifluoromethyl)aniline
Supplier: COMBI-BLOCKS
Certificates
About this item
CAS Number:393-36-2
:
Formula:C₇H₅BrF₃N
MDL Number:MFCD00007827
:240.02 g/mol
Boiling Pt:81…84 °C (0.5 mmHg)
Melting Pt:55…56 °C
EINECS:206-885-1
UN:2811
ADR:6.1,III
Frequently Bought Together