Order Entry
Germany
ContactUsLinkComponent
 
Pesticide reference standard, 2-Chloroaniline 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA11350000AL
Supplier:  EHRENSTORFER
CAS Number:  
Pesticide reference standard, 2-Chloroaniline 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA11350000AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    95-51-2
  • Description:
    2-Chloroaniline 100 µg/ml in acetonitrile
  • Formula:
    C₆H₆ClN
  • Description:
    2-Chloroaniline

Specifications

About this item