Order Entry
Germany
ContactUsLinkComponent
 
Pesticide reference standard, 2,4,5-Trichloroaniline 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL17700200CY
 :  
Pesticide reference standard, 2,4,5-Trichloroaniline 10 µg/ml in cyclohexane, Dr Ehrenstorfer
  EHERL17700200CY
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    636-30-6
  • Description:
    2,4,5-Trichloroaniline 10 µg/ml in cyclohexane
  • Formula:
    C₆H₄Cl₃N
  • Description:
    2,4,5-Trichloroaniline