Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dexamethasone 21-acetate
Dexamethasone-21-acetate, Sigma-Aldrich®
Dexamethasone-21-acetate, Sigma-Aldrich®
Dexamethasone-21-acetate, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Dexamethasone-21-acetate, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
1177-87-3
:
Formula:
C₂₄H₃₁FO₆
MDL Number:
MFCD00027407
:
434.5 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
214-646-8