1,4-Diaminobutane dihydrochloride
Catalog # MOLE25523156-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Density:1.31 g/cm³ (20 °C)
:161.07 g/mol
EINECS:206-375-9
CAS Number:333-93-7
Formula:C₄H₁₄Cl₂N₂
MDL Number:MFCD00012526