Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Bis-trispropane ≥98%

Bis-trispropane ≥98%
Certificates
Bis-trispropane ≥98%
Bis-trispropane ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 64431-96-5 Molecular Formula: CH2[CH2NHC(CH2OH)3]2 Formula Weight: 282.34 Physical Form: Powder Melting Point: 168-170° MDL No.: MFCD00004689

Synonyms:  1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
  • CAS Number:64431-96-5
  • Formula:C₁₁H₂₆N₂O₆
  • MDL Number:MFCD00004689
  • Molecular Weight:282.34 g/mol
  • Melting Pt:168…170 °C
  • Storage Temperature:Ambient
  • EINECS:264-899-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 59,90 €
    59,90 €
    1 * 25 g
       59,90 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 194,00 €
    194,00 €
    1 * 100 g
       194,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 713,00 €
    713,00 €
    1 * 500 g
       713,00 €