Order Entry
Germany
ContactUsLinkComponent
Acetonitrile, Uvasol® for spectroscopy, Supelco®
Acetonitrile, Uvasol® for spectroscopy, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
undefined
Acetonitrile, Uvasol® for spectroscopy, Supelco®
Supplier:  Merck
   Sustainable Options Available

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -45.7 °C
  • Molar Mass:
    41.05
  • Boiling Point:
    81.6 °C
  • Vapor Pressure:
    97 hPA
  • Flash Point:
    2 °C
  • Refractive Index:
    1.34
  • Explosion Limit:
    3.0 - 17%
  • Density:
    0.786 g/cm³
  • pH:
    (H₂0) N/A
  • Ignition Point:
    524 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

Related Information