Order Entry
Germany
ContactUsLinkComponent
3,3'-Diaminobenzidine (DAB) 97% (by HPLC), Sigma-Aldrich®
3,3'-Diaminobenzidine (DAB) 97% (by HPLC), Sigma-Aldrich®
Certificates
undefined
3,3'-Diaminobenzidine (DAB) 97% (by HPLC), Sigma-Aldrich®

Specification Test Results

  • Appearance (colour):
    Off white to yellow to brown
  • Appearance (form):
    Powder
  • Purity (HPLC area %):
    ≥97%
  • Infrared spectrum:
    Conforms to structure

Specification Test Results